missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methoxyethoxy)benzenboronsäure, 95%, Thermo Scientific™

Catalog Number 15411079
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
265664-52-6
C9H13BO4
196.01
MFCD03452784
ODKMFCPXFMQWHA-UHFFFAOYSA-N
4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy phenylboronic acid, 4-2-methoxyethoxy benzeneboronic acid, acmc-209gsb, 4-2-methoxyethoxy phenylboronicacid, boronic acid, 4-2-methoxyethoxy phenyl
23508303
[4-(2-methoxyethoxy)phenyl]boronic acid
COCCOC1=CC=C(C=C1)B(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
265664-52-6
C9H13BO4
196.01
MFCD03452784
ODKMFCPXFMQWHA-UHFFFAOYSA-N
4-2-methoxyethoxy phenyl boronic acid, 4-2-methoxyethoxy phenylboronic acid, 4-2-methoxyethoxy benzeneboronic acid, acmc-209gsb, 4-2-methoxyethoxy phenylboronicacid, boronic acid, 4-2-methoxyethoxy phenyl
23508303
[4-(2-methoxyethoxy)phenyl]boronic acid
COCCOC1=CC=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(2-Methoxyethoxy)benzeneboronic acid
Menge 1 g
Formelmasse 196.01
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'