missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 4-(3 -Hydroxypropyl)benzenboronsäure, 96 %
Click to view available options
Menge:
250 mg
Spezifikation
Chemischer Name oder Material | 4-(3-Hydroxypropyl)benzeneboronic acid |
CAS | 850568-48-8 |
Schmelzpunkt | 70°C to 74°C |
Summenformel | C9H13BO3 |
MDL-Nummer | MFCD04973734 |
Menge | 250 mg |
Synonym | 4-3-hydroxypropyl phenylboronic acid, 4-3-hydroxypropyl benzeneboronic acid, 4-3-hydroxypropyl phenyl boronic acid, boronic acid, 4-3-hydroxypropyl phenyl, boronic acid,b-4-3-hydroxypropyl phenyl |
InChI-Schlüssel | KCRVWMBFIDPDIF-UHFFFAOYSA-N |
SMILES | B(C1=CC=C(C=C1)CCCO)(O)O |
IUPAC-Name | [4-(3-Hydroxypropyl)phenyl]boronsäure |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'