Learn More
4,4,5,5-Tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane, 97%, BLD Pharm™
Product Code. 30948521
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
100 g
500 g
Unit Size:
100g
1g
250mg
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948521 1 g 1g
30945537 250 mg 250mg
30938146 100 g 100g
30947838 500 g 500g
30945129 5 g 5g
30931884 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30948521 Supplier BLD Pharm Supplier No. BD1432721g
 Restricted Item
Verfügbar ab: 27-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
CAS 256652-04-7
Molecular Formula C16H19BO2
MDL Number MFCD05663868
Quantity 1 g
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=CC=C2)C=C1
Molecular Weight (g/mol) 254.13
Percent Purity 97%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane