missing translation for 'onlineSavingsMsg'
Learn More

4-(4,6-Dichloro-2 -pyrimidinyl)Morpholin, 97 %, Thermo Scientific™

Catalog Number 15480528
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
10397-13-4
C8H9Cl2N3O
234.08
MFCD05022359
OXCOCPRVQUEIOL-UHFFFAOYSA-N
4-4,6-dichloropyrimidin-2-yl morpholine, 4-4,6-dichloro-pyrimidin-2-yl-morpholine, 4-4,6-dichloro-2-pyrimidyl morpholine, 2-morpholino-4,6-dichloropyrimidine, 4,6-dichloro-2-morpholinopyrimidine, 2-4-morpholino-4,6-dichloropyrimidine, morpholine,4-4,6-dichloro-2-pyrimidinyl, morpholine, 4-4,6-dichloro-2-pyrimidinyl, 4-4,6-dichloro-2-pyrimidinyl morpholine, pubchem21502
2772068
4-(4,6-Dichlorpyrimidin-2-yl)morpholin
ClC1=CC(Cl)=NC(=N1)N1CCOCC1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
10397-13-4
C8H9Cl2N3O
234.08
MFCD05022359
OXCOCPRVQUEIOL-UHFFFAOYSA-N
4-4,6-dichloropyrimidin-2-yl morpholine, 4-4,6-dichloro-pyrimidin-2-yl-morpholine, 4-4,6-dichloro-2-pyrimidyl morpholine, 2-morpholino-4,6-dichloropyrimidine, 4,6-dichloro-2-morpholinopyrimidine, 2-4-morpholino-4,6-dichloropyrimidine, morpholine,4-4,6-dichloro-2-pyrimidinyl, morpholine, 4-4,6-dichloro-2-pyrimidinyl, 4-4,6-dichloro-2-pyrimidinyl morpholine, pubchem21502
2772068
4-(4,6-Dichlorpyrimidin-2-yl)morpholin
ClC1=CC(Cl)=NC(=N1)N1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4,6-Dichloro-2-pyrimidinyl)morpholine
Menge 1 g
Formelmasse 234.08
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'