missing translation for 'onlineSavingsMsg'
Learn More

4-(4'-Chlor-4-Biphenylsulfonylamino)Benzoesäure, 96 %, Thermo Scientific™

Catalog Number p-7032856
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885269-32-9
C19H14ClNO4S
387.834
MFCD06409447
AYWSYJTWHIPSFL-UHFFFAOYSA-N
4-4'-chloro-4-biphenylylsulfonylamino benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-ylsulfonamido benzoic acid
44669160
4-[[4-(4-chlorphenyl)phenyl]sulfonylamino]benzoesäure
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885269-32-9
C19H14ClNO4S
387.834
MFCD06409447
AYWSYJTWHIPSFL-UHFFFAOYSA-N
4-4'-chloro-4-biphenylylsulfonylamino benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-sulfonamido benzoic acid, 4-4'-chloro-1,1'-biphenyl-4-ylsulfonamido benzoic acid
44669160
4-[[4-(4-chlorphenyl)phenyl]sulfonylamino]benzoesäure
C1=CC(=CC=C1C2=CC=C(C=C2)Cl)S(=O)(=O)NC3=CC=C(C=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4'-Chloro-4-biphenylylsulfonylamino)benzoic acid
Schmelzpunkt 298°C
Menge 250 mg
Formelmasse 387.84
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'