missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Cyanphenyl)-2-methylthiazol, 97 %, Thermo Scientific™

Catalog Number p-7036218
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
127406-79-5
C11H8N2S
200.26
MFCD01765038
FOKBMJMDQCPNJI-UHFFFAOYSA-N
716353
4-(2-Methyl-1,3-Thiazol-4-yl)Benzonitril
CC1=NC(=CS1)C1=CC=C(C=C1)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
127406-79-5
C11H8N2S
200.26
MFCD01765038
FOKBMJMDQCPNJI-UHFFFAOYSA-N
716353
4-(2-Methyl-1,3-Thiazol-4-yl)Benzonitril
CC1=NC(=CS1)C1=CC=C(C=C1)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4-Cyanophenyl)-2-methylthiazole
Menge 250 mg
Synonym 4-2-methyl-1,3-thiazol-4-yl benzonitrile, 4-2-methylthiazol-4-yl benzonitrile, benzonitrile,4-2-methyl-4-thiazolyl, acmc-20msfj, maybridge3_006920, 4-4-cyanophenyl-2-methylthiazole, 4-2-methylthiazol-4-yl-benzonitrile, benzonitrile, 4-2-methyl-4-thiazolyl, 4-2-methyl-1,3-thiazol-4-yl benznitrile, 4-2-methyl-1,3-thiazol-4-yl benzenecarbonitrile
Formelmasse 200.26
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'