missing translation for 'onlineSavingsMsg'
Learn More

4,4-Difluorcyclohexancarbonsäure, 98 %, Thermo Scientific Chemicals

Catalog Number p-7028490
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122665-97-8
C7H10F2O2
164.152
MFCD03788493
HYIUDFLDFSIXTR-UHFFFAOYSA-N
4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
2779136
4,4-Difluorcyclohexan-1-Carbonsäure
C1CC(CCC1C(=O)O)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
122665-97-8
C7H10F2O2
164.152
MFCD03788493
HYIUDFLDFSIXTR-UHFFFAOYSA-N
4,4-difluorocyclohexanecarboxylic acid, cyclohexanecarboxylic acid, 4,4-difluoro, 4,4-difluorocyclohexylcarboxylic acid, 4,4-difluorocyclohexanoic acid, 4, 4-difluoro-cyclohexane carboxylic acid, 4,4-difluorocyclohexane carboxylic acid, 4,4-difluorocyclohexanecarboxylicacid, pubchem15915, acmc-209aln, ksc174k3t
2779136
4,4-Difluorcyclohexan-1-Carbonsäure
C1CC(CCC1C(=O)O)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4,4-Difluorocyclohexanecarboxylic acid
Schmelzpunkt 104°C to 107°C
Menge 250 mg
Formelmasse 164.15
Reinheit (%) 98%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'