missing translation for 'onlineSavingsMsg'
Learn More

4,4-Dimethylcyclohexa-1,5-dien-1-Boronsäuremononatriumsalz, 96 %

Catalog Number 15480106
Click to view available options
Menge:
250 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4,4-Dimethylcyclohexa-1,5-diene-1-boronic acid monosodium salt
CAS 871329-70-3
Schmelzpunkt 300°C to 302°C
Summenformel C8H12BNaO2
MDL-Nummer MFCD16036162
Menge 250 mg
Synonym 4,4-dimethylcyclohexa-1,5-dienylboronic acid monosodium salt, 4,4-dimethylcyclohexa-1,5-dienylboronic acid, monosodium salt, sodium hydrogen 4,4-dimethylcyclohexa-1,5-dien-1-ylboronate, sodium hydrogen 4,4-dimethylcyclohexa-1,5-dien-1-yl boronate, 4,4-dimethyl cyclohexa-1,5-dienylboronic acid sodium salt, 4,4-dimethylcyclohexa-15-dienylboronic acid monosodium salt, sodium hydrogen-4,4-dimethylcyclohexa-1,5-dienylboronate, 4,4-dimethylcyclohexa-1,5-dienyl boronic acid monosodium salt, 4,4-dimethylcyclohexa-1,5-diene-1-boronic acid monosodium salt
InChI-Schlüssel YBUXCAFVTNSKLJ-UHFFFAOYSA-N
SMILES B(C1=CCC(C=C1)(C)C)(O)[O-].[Na+]
IUPAC-Name Natrium;(4,4-dimethylcyclohexa-1,5-dien-1-yl)-hydroxyborinat
Molekulargewicht (g/mol) 173.982
PubChem CID 44119640
Formelmasse 173.98
Reinheit (%) 96%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'