missing translation for 'onlineSavingsMsg'
Learn More

4-(4'-Methoxy-4-biphenylsulfonylaminomethyl)benzoesäure, 96 %, Thermo Scientific™

Catalog Number p-7031274
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885269-44-3
C21H19NO5S
397.445
MFCD06409459
QVOYNMQWWYYZBY-UHFFFAOYSA-N
4-4'-methoxy-1,1'-biphenyl-4-sulfonamido methyl benzoic acid, 4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido methyl benzoic acid, 4-4'-methoxy-4-biphenylylsulfonylaminomethyl benzoic acid, 4-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino methyl benzoic acid
44669169
4-[[[4[-(4-methoxyphenyl)phenyl]sulfonylamino]methyl]Benzoesäure
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NCC3=CC=C(C=C3)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
885269-44-3
C21H19NO5S
397.445
MFCD06409459
QVOYNMQWWYYZBY-UHFFFAOYSA-N
4-4'-methoxy-1,1'-biphenyl-4-sulfonamido methyl benzoic acid, 4-4'-methoxy-1,1'-biphenyl-4-ylsulfonamido methyl benzoic acid, 4-4'-methoxy-4-biphenylylsulfonylaminomethyl benzoic acid, 4-4'-methoxy 1,1'-biphenyl-4-sulfonyl amino methyl benzoic acid
44669169
4-[[[4[-(4-methoxyphenyl)phenyl]sulfonylamino]methyl]Benzoesäure
COC1=CC=C(C=C1)C2=CC=C(C=C2)S(=O)(=O)NCC3=CC=C(C=C3)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4'-Methoxy-4-biphenylylsulfonylaminomethyl)benzoic acid
Schmelzpunkt >250°C
Menge 250 mg
Formelmasse 397.45
Reinheit (%) 96%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'