missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methoxyphenylethynyl)-benzolboronsäurepinakolester , 95 %, Thermo Scientific™

Catalog Number p-7034246
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1190376-24-9
C21H23BO3
334.22
MFCD16294533
RRYKLXOOBFDYDY-UHFFFAOYSA-N
2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, 2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb224, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
71306524
COC1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1190376-24-9
C21H23BO3
334.22
MFCD16294533
RRYKLXOOBFDYDY-UHFFFAOYSA-N
2-4-4-methoxy-phenylethynyl-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 4-4-methoxyphenylethynyl benzeneboronic acid pinacol ester, 2-4-2-4-methoxyphenyl ethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, amtb224, 2-4-4-methoxyphenylethynyl phenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
71306524
COC1=CC=C(C=C1)C#CC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4-Methoxyphenylethynyl)benzeneboronic acid pinacol ester
Menge 1 g
IUPAC-Name 2-[4-[2-(4-Methoxyphenyl)Ethynyl]Phenyl]-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan
Formelmasse 334.22
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'