missing translation for 'onlineSavingsMsg'
Learn More

4-(4-Methyl-1-Piperazinylcarbonyl)benzolboronsäure-Hydrochlorid, 95 %, Thermo Scientific™

Catalog Number p-7035577
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
913835-43-5
C12H19BClN3O3
299.56
MFCD09027217
DPTCRDJSYQTDGH-UHFFFAOYSA-N
4-4-methylpiperazin-1-yl carbamoyl phenyl boronic acid hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl phenylboronic acid hydrochloride, c12h17bn2o3.clh, n-4-methylpiperazinyl 4-boronobenzamide hcl, n4-methylpiperazinyl 4-boronobenzamide hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl-phenyl boronic acid hydrochloride
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
913835-43-5
C12H19BClN3O3
299.56
MFCD09027217
DPTCRDJSYQTDGH-UHFFFAOYSA-N
4-4-methylpiperazin-1-yl carbamoyl phenyl boronic acid hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl phenylboronic acid hydrochloride, c12h17bn2o3.clh, n-4-methylpiperazinyl 4-boronobenzamide hcl, n4-methylpiperazinyl 4-boronobenzamide hydrochloride, 4-4-methylpiperazin-1-yl carbamoyl-phenyl boronic acid hydrochloride

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(4-Methyl-1-piperazinylcarbonyl)benzeneboronic acid hydrochloride
Schmelzpunkt 187°C to 189°C
Menge 250 mg
Formelmasse 284.55
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'