missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dicyan-2-(3-methylphenyl)-imidazol, 97 %, Thermo Scientific™

Catalog Number p-7036049
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-10-4
C12H8N4
208.224
MFCD20265228
OIQBYNHKZGZUMW-UHFFFAOYSA-N
4,5-dicyano-2-3-methylphenyl imidazole, 2-3-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-m-tolyl imidazole
73995740
2-(3-Methylphenyl)-1H-Imidazol-4,5-Dicarbonitril
CC1=CC=CC(=C1)C2=NC(=C(N2)C#N)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1347815-10-4
C12H8N4
208.224
MFCD20265228
OIQBYNHKZGZUMW-UHFFFAOYSA-N
4,5-dicyano-2-3-methylphenyl imidazole, 2-3-methylphenyl-1h-imidazole-4,5-dicarbonitrile, 4,5-dicyano-2-m-tolyl imidazole
73995740
2-(3-Methylphenyl)-1H-Imidazol-4,5-Dicarbonitril
CC1=CC=CC(=C1)C2=NC(=C(N2)C#N)C#N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4,5-Dicyano-2-(3-methylphenyl)imidazole
Menge 250 mg
Formelmasse 208.22
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'