Learn More
Thermo Scientific Chemicals 4,6-O-Ethyliden-D-glucopyranose, 90 %
Catalog Number p-7043252
Click to view available options
Menge:
1 g
5 g
13224-99-2
C8H14O6
206.19
MFCD00006820
VZPBLPQAMPVTFO-UHFFFAOYNA-N
4,6-o-ethylidene-a-d-glucose
21581900
(2R,4aR,6S,7R,8R,8aS)-2-Methyl-4,4a,6,7,8,8a-Hexahydropyrano[3,2-d][1,3]Dioxin-6,7,8-Triol
CC1OCC2OC(O)C(O)C(O)C2O1
13224-99-2
C8H14O6
206.19
MFCD00006820
VZPBLPQAMPVTFO-UHFFFAOYNA-N
4,6-o-ethylidene-a-d-glucose
21581900
(2R,4aR,6S,7R,8R,8aS)-2-Methyl-4,4a,6,7,8,8a-Hexahydropyrano[3,2-d][1,3]Dioxin-6,7,8-Triol
CC1OCC2OC(O)C(O)C(O)C2O1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4,6-O-Ethylidene-D-glucopyranose
Farbe Weiß
Flammpunkt >110°C (230°F)
Menge 1 g
Formelmasse 206.19
Reinheit (%) 90%
Physikalische Form Pulver