missing translation for 'onlineSavingsMsg'
Learn More

4-(9-Carbazolylmethyl)-benzolboronsäurepinakolester, 95 %, Thermo Scientific™

Catalog Number p-7034598
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1315281-51-6
C25H26BNO2
383.298
MFCD18827487
AKZHQKVQCWCVBW-UHFFFAOYSA-N
9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-9h-carbazole, 4-9-carbazolylmethyl benzeneboronic acid pinacol ester, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbazole, amtb382, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-9h-carbazole
71306542
9-[4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Methyl]Carbazol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3C4=CC=CC=C4C5=CC=CC=C53
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1315281-51-6
C25H26BNO2
383.298
MFCD18827487
AKZHQKVQCWCVBW-UHFFFAOYSA-N
9-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-9h-carbazole, 4-9-carbazolylmethyl benzeneboronic acid pinacol ester, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzyl-9h-carbazole, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbazole, amtb382, 9-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl-9h-carbazole
71306542
9-[4-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Phenyl]Methyl]Carbazol
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN3C4=CC=CC=C4C5=CC=CC=C53

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(9-Carbazolylmethyl)benzeneboronic acid pinacol ester
Menge 1 g
Formelmasse 383.3
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'