Learn More
4-Amino-1-isobutylpiperidin, 95 %, Thermo Scientific™
Catalog Number p-7033753
Click to view available options
Menge:
250 mg
1 g
172478-05-6
C9H18N2O
170.256
MFCD08061057
LULDRPDOMFMBIC-UHFFFAOYSA-N
1-4-aminopiperidin-1-yl-2-methylpropan-1-one, 1-isobutyrylpiperidin-4-amine, 4-amino-1-isobutyrylpiperidine, 1-propanone, 1-4-amino-1-piperidinyl-2-methyl, 1-4-aminopiperidyl-2-methylpropan-1-one, acmc-1bz3p, 1-isobutyryl-4-piperidinamine, 4-piperidinamine, 1-2-methyl-1-oxopropyl
16772991
1-(4-Aminopiperidin-1-yl)-2-Methylpropan-1-on
CC(C)C(=O)N1CCC(CC1)N
172478-05-6
C9H18N2O
170.256
MFCD08061057
LULDRPDOMFMBIC-UHFFFAOYSA-N
1-4-aminopiperidin-1-yl-2-methylpropan-1-one, 1-isobutyrylpiperidin-4-amine, 4-amino-1-isobutyrylpiperidine, 1-propanone, 1-4-amino-1-piperidinyl-2-methyl, 1-4-aminopiperidyl-2-methylpropan-1-one, acmc-1bz3p, 1-isobutyryl-4-piperidinamine, 4-piperidinamine, 1-2-methyl-1-oxopropyl
16772991
1-(4-Aminopiperidin-1-yl)-2-Methylpropan-1-on
CC(C)C(=O)N1CCC(CC1)N
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Amino-1-isobutyrylpiperidine
Menge 250 mg
Formelmasse 170.26
Reinheit (%) 95%