missing translation for 'onlineSavingsMsg'
Learn More

4-(Aminomethyl)benzenboronsäure-Pinacol Ester-Hydrochlorid, 95 %, Thermo Scientific Chemicals

Catalog Number p-7034989
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850568-55-7
C13H21BClNO2
269.576
MFCD02179455
KPECMJIHZZWTJN-UHFFFAOYSA-N
4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl
16427088
[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamin;hydrochlorid
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850568-55-7
C13H21BClNO2
269.576
MFCD02179455
KPECMJIHZZWTJN-UHFFFAOYSA-N
4-aminomethylphenylboronic acid, pinacol ester, hcl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hydrochloride, 4-aminomethylphenylboronic acid pinacol ester hcl, 4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hydrochloride, benzenemethanamine,4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylamine hydrochloride, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanamine hcl, 4-aminomethylohenylboronic acid pinacl ester hcl
16427088
[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methanamin;hydrochlorid
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)CN.Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Aminomethyl)benzeneboronic acid pinacol ester hydrochloride
Schmelzpunkt 228°C to 238°C
Menge 250 mg
Formelmasse 269.58
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'