Learn More
4-Benzofurazancarboxaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 4-Benzofurazancarboxaldehyde |
| CAS | 32863-32-4 |
| Molecular Formula | C7 H4 N2 O2 |
| Quantity | 100 mg |
| Synonym | 2,1,3-Benzoxadiazole-4-Carbaldehyde |
| InChI Formula | InChI=1S/C7H4N2O2/c10-4-5-2-1-3-6-7(5)9-11-8-6/h1-4H |
| SMILES | O=Cc1cccc2nonc12 |
| IUPAC Name | 2,1,3-benzoxadiazole-4-carbaldehyde |
| Molecular Weight (g/mol) | 148.12 |
| Formula Weight | 148.0273 |