missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2-Chlor-5-Hydroxypyrimidin, 95 %, Thermo Scientific™

Catalog Number p-7029522
Click to view available options
Menge:
250 mg
1 g
Este artículo no se puede devolver. Vea la política de devoluciones
885952-28-3
C11H9ClN2O2
236.66
MFCD06798283
URJRKXXIIWYWIS-UHFFFAOYSA-N
4-benzyloxy-2-chloropyrimidin-5-ol, 4-benzyloxy-2-chloro-5-hydroxypyrimidine, 4-benzyloxy-2-chloro-5-pyrimidinol, 5-pyrimidinol,2-chloro-4-phenylmethoxy
42552982
2-Chlor-4-Phenylmethoxypyrimidin-5-ol
OC1=CN=C(Cl)N=C1OCC1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
885952-28-3
C11H9ClN2O2
236.66
MFCD06798283
URJRKXXIIWYWIS-UHFFFAOYSA-N
4-benzyloxy-2-chloropyrimidin-5-ol, 4-benzyloxy-2-chloro-5-hydroxypyrimidine, 4-benzyloxy-2-chloro-5-pyrimidinol, 5-pyrimidinol,2-chloro-4-phenylmethoxy
42552982
2-Chlor-4-Phenylmethoxypyrimidin-5-ol
OC1=CN=C(Cl)N=C1OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Benzyloxy-2-chloro-5-hydroxypyrimidine
Menge 250 mg
Formelmasse 236.66
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'