missing translation for 'onlineSavingsMsg'
Learn More

4-[Bis(2-methoxy-2-oxoethyl)amino]-3-[2-[2-[bis(2-methoxy-2-oxoethyl)amino]-5-methylphenoxy]ethoxy]-benzoesäure, TRC

Product Code. 30341038
Change view
Click to view available options
Menge:
25 mg
Unit Size:
25 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30341038 25 mg 25 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30341038 Supplier Toronto Research Chemicals Supplier No. TRCB41414525MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 4-[Bis(2-methoxy-2-oxoethyl)amino]-3-[2-[2-[bis(2-methoxy-2-oxoethyl)amino]-5-methylphenoxy]ethoxy]-benzoic Acid
CAS 773871-94-6
Summenformel C28H34N2O12
Menge 25 mg
Synonym 4-(Bis(2-methoxy-2-oxoethyl)amino)-3-(2-(2-(bis(2-methoxy-2-oxoethyl)amino)-5-methylphenoxy)ethoxy)benzoic Acid
InChI-Formel InChI=1S/C28H34N2O12/c1-18-6-8-20(29(14-24(31)37-2)15-25(32)38-3)22(12-18)41-10-11-42-23-13-19(28(35)36)7-9-21(23)30(16-26(33)39-4)17-27(34)40-5/h6-9,12-13H,10-11,14-17H2,1-5H3,(H,35,36)
SMILES COC(=O)CN(CC(=O)OC)c1ccc(C)cc1OCCOc2cc(ccc2N(CC(=O)OC)CC(=O)OC)C(=O)O
IUPAC-Name 4-[Bis(2-methoxy-2-oxoethyl)amino]-3-[2-[2-[Bis(2-methoxy-2-oxoethyl)amino]-5-methylphenoxy]ethoxy]benzoesäure
Molekulargewicht (g/mol) 590.58
Formelmasse 590.2112
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.