missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)-2-Chlorbenzoesäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7035081
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
232275-73-9
C12H14ClNO4
271.697
MFCD02682164
HDYBNYDKCZJPPX-UHFFFAOYSA-N
n-boc-4-amino-2-chlorobenzoic acid, 4-tert-butoxycarbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino-benzoic acid, 4-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino benzoic acid, 4-tert-butoxycarbonylamino-2-chloro-benzoic acid, 4-tert-butoxycarbonylamino-2-chlorobenzoic acid, benzoic acid,2-chloro-4-1,1-dimethylethoxy carbonyl amino
17750449
2-chlor-4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoesäure
CC(C)(C)OC(=O)NC1=CC(=C(C=C1)C(=O)O)Cl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
232275-73-9
C12H14ClNO4
271.697
MFCD02682164
HDYBNYDKCZJPPX-UHFFFAOYSA-N
n-boc-4-amino-2-chlorobenzoic acid, 4-tert-butoxycarbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino-benzoic acid, 4-tert-butoxy carbonyl amino-2-chlorobenzoic acid, 2-chloro-4-boc-amino benzoic acid, 4-tert-butoxycarbonylamino-2-chloro-benzoic acid, 4-tert-butoxycarbonylamino-2-chlorobenzoic acid, benzoic acid,2-chloro-4-1,1-dimethylethoxy carbonyl amino
17750449
2-chlor-4-[(2-methylpropan-2-yl)oxycarbonylamino]benzoesäure
CC(C)(C)OC(=O)NC1=CC(=C(C=C1)C(=O)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Boc-amino)-2-chlorobenzoic acid
Menge 250 mg
Formelmasse 271.7
Reinheit (%) 97%

RUO – Nur für Forschungszwecke

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'