missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-amino)benzenboronsäure-Pinacol ester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7047709
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
330793-01-6
C17H26BNO4
319.21
MFCD02179439
HSJNIOYPTSKQBD-UHFFFAOYSA-N
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
2734617
tert-Butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamat
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
330793-01-6
C17H26BNO4
319.21
MFCD02179439
HSJNIOYPTSKQBD-UHFFFAOYSA-N
4-n-boc-amino phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino benzeneboronic acid pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 4-boc-amino phenylboronic acid pinacol cyclic ester, 4-boc-aminophenylboronic acid, pinacol ester, 4-tert-butoxycarbonyl aminophenylboronic acid, pinacol ester, tert-butyl n-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenylcarbamate
2734617
tert-Butyl N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamat
CC(C)(C)OC(=O)NC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Boc-amino)benzeneboronic acid pinacol ester
Schmelzpunkt 169°C to 170°C
Menge 1 g
Formelmasse 319.21
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'