missing translation for 'onlineSavingsMsg'
Learn More

4'-Brom-2'-Fluoracetanilid, 98 %, Thermo Scientific™

Catalog Number p-7022806
Click to view available options
Menge:
10 g
50 g
250 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
326-66-9
C8H7BrFNO
232.052
MFCD00042588
BCYGKMDWQBWUSC-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl acetamide, 4'-bromo-2'-fluoroacetanilide, 4-bromo-2-fluoroacetanilide, acetamide, n-4-bromo-2-fluorophenyl, pubchem14886, acmc-1cooc, ksc497m2b, timtec-bb sbb009911, attercop-chm at130693
67593
N-(4-Brom-2-fluorphenyl)acetamid
CC(=O)NC1=C(C=C(C=C1)Br)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
326-66-9
C8H7BrFNO
232.052
MFCD00042588
BCYGKMDWQBWUSC-UHFFFAOYSA-N
n-4-bromo-2-fluorophenyl acetamide, 4'-bromo-2'-fluoroacetanilide, 4-bromo-2-fluoroacetanilide, acetamide, n-4-bromo-2-fluorophenyl, pubchem14886, acmc-1cooc, ksc497m2b, timtec-bb sbb009911, attercop-chm at130693
67593
N-(4-Brom-2-fluorphenyl)acetamid
CC(=O)NC1=C(C=C(C=C1)Br)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4'-Bromo-2'-fluoroacetanilide
Schmelzpunkt 154°C to 158°C
Menge 10 g
Beilstein 3250459
Formelmasse 232.05
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'