missing translation for 'onlineSavingsMsg'
Learn More

4-Carbamoylbenzolboronsäure, 98 %, Thermo Scientific™

Catalog Number p-7035626
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
123088-59-5
C7H8BNO3
164.96
MFCD03411940
GNRHNKBJNUVWFZ-UHFFFAOYSA-N
4-aminocarbonylphenylboronic acid, 4-carbamoylphenyl boronic acid, benzamide-4-boronic acid, 4-aminocarbonyl benzeneboronic acid, 4-carbamoylbenzeneboronic acid, 4-aminocarbonylphenyl boronic acid, 4-dihydroxyboranyl benzamide, p-aminocarbonyl phenylboronic acid, boronic acid, 4-aminocarbonyl phenyl, 4-aminocarbonyl phenylboronic acid
2737811
(4-carbamoylphenyl)boronic acid
NC(=O)C1=CC=C(C=C1)B(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
123088-59-5
C7H8BNO3
164.96
MFCD03411940
GNRHNKBJNUVWFZ-UHFFFAOYSA-N
4-aminocarbonylphenylboronic acid, 4-carbamoylphenyl boronic acid, benzamide-4-boronic acid, 4-aminocarbonyl benzeneboronic acid, 4-carbamoylbenzeneboronic acid, 4-aminocarbonylphenyl boronic acid, 4-dihydroxyboranyl benzamide, p-aminocarbonyl phenylboronic acid, boronic acid, 4-aminocarbonyl phenyl, 4-aminocarbonyl phenylboronic acid
2737811
(4-carbamoylphenyl)boronic acid
NC(=O)C1=CC=C(C=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Carbamoylbenzeneboronic acid
Schmelzpunkt 242°C to 252°C
Menge 1 g
Formelmasse 164.96
Reinheit (%) 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'