missing translation for 'onlineSavingsMsg'
Learn More

4-Chlor-2,5-difluorbenzoesäure, 98 %, Thermo Scientific™

Catalog Number 15461808
Click to view available options
Menge:
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132794-07-1
C7H2ClF2O2
191.54
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-M
2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
2733274
4-Chlor-2,5-difluorbenzoesäure
[O-]C(=O)C1=CC(F)=C(Cl)C=C1F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
132794-07-1
C7H2ClF2O2
191.54
MFCD00077480
PEPCYJSDHYMIFN-UHFFFAOYSA-M
2,5-difluoro-4-chlorobenzoic acid, 4-chloro-2,5-difluorobenzoicacid, 4-chloro-2,5-difluoro-benzoic acid, benzoic acid, 4-chloro-2,5-difluoro, pubchem1367, intermediates-zcf02018, acmc-1c2s0, 2,5-difluoro-4-chlorobenzoicacid, attercop-chm at136451, 2,5-difluoro-4-chloro-benzoic acid
2733274
4-Chlor-2,5-difluorbenzoesäure
[O-]C(=O)C1=CC(F)=C(Cl)C=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-2,5-difluorobenzoic acid
Schmelzpunkt 154°C to 157°C
Menge 25 g
Formelmasse 192.55
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'