Learn More
4-Chlor-2-Fluorbenzenboronsäure, 97 %, Thermo Scientific Chemicals
Catalog Number p-7030461
Click to view available options
Menge:
1 g
10 g
160591-91-3
C6H5BClFO2
174.362
MFCD02684293
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
4193595
(4-Chlor-2-Fluorphenyl)boronsäure
B(C1=C(C=C(C=C1)Cl)F)(O)O
160591-91-3
C6H5BClFO2
174.362
MFCD02684293
YBNDRTRLXPEWKQ-UHFFFAOYSA-N
4-chloro-2-fluorobenzeneboronic acid, 4-chloro-2-fluorophenyl boronic acid, 4-chloro-2-fluorophenylboronicacid, 2-fluoro-4-chlorophenylboronic acid, b-4-chloro-2-fluorophenyl-boronic acid, 4-chloro-2-fluorophenyl-boronic acid, boronic acid, 4-chloro-2-fluorophenyl, boronic acid, b-4-chloro-2-fluorophenyl, pubchem5034, 4-chloro-2-fluoro-phenyl boronic acid
4193595
(4-Chlor-2-Fluorphenyl)boronsäure
B(C1=C(C=C(C=C1)Cl)F)(O)O
Used as intermediates for pharmaceutical and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Anwendungen
Wird als Zwischenprodukte für Pharmazeutika und Agrochemikalien verwendet.
Löslichkeit
Unlöslich in Wasser.
Hinweise
Kühl lagern. Behälter an einem trockenen und gut belüfteten Ort dicht verschlossen halten. Von Oxidationsmitteln entfernt aufbewahren.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-2-fluorobenzeneboronic acid
Schmelzpunkt 246°C to 250°C
Menge 1 g
Löslichkeitsinformationen Insoluble in water.
Formelmasse 174.37
Reinheit (%) 97%