missing translation for 'onlineSavingsMsg'
Learn More

4-Chlor-3-(Trifluormethyl)pyridinhydrochlorid, 97 %, Thermo Scientific Chemicals

Catalog Number p-7047646
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
732306-24-0
C6H4Cl2F3N
218.00
MFCD04972770
HCKXJUZITVCDND-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
2782815
4-Chlor-3-(trifluormethyl)pyridinhydrochlorid
Cl.FC(F)(F)C1=C(Cl)C=CN=C1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
732306-24-0
C6H4Cl2F3N
218.00
MFCD04972770
HCKXJUZITVCDND-UHFFFAOYSA-N
4-chloro-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethyl pyridinehydrochloride, 4-chloro-3-trifluoromethyl pyridine, chloride, 4-chloro-3-trifluoromethyl pyridine hcl, 4-chloro-3-trifluoromethylpyridine hydrochloride, 3-chloro-4-trifluoromethyl pyridine hydrochlrorid, 4-chloranyl-3-trifluoromethyl pyridine hydrochloride, 4-chloro-3-trifluoromethylpyridine hydrochloride salt
2782815
4-Chlor-3-(trifluormethyl)pyridinhydrochlorid
Cl.FC(F)(F)C1=C(Cl)C=CN=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
Schmelzpunkt ∼135°C (sublimation)
Menge 250 mg
Formelmasse 218.01
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'