missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-N-(2-ethylphenyl)butyramid, 97 %, Thermo Scientific™

Catalog Number p-7040108
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
42276-57-3
C12H16ClNO
225.716
MFCD03362124
VAWRRNXJFXHSSI-UHFFFAOYSA-N
4-chloro-n-2-ethylphenyl butanamide, 4-chloro-n-2-ethylphenyl butyramide
3523909
4-Chlor-N-(2-ethylphenyl)butanamid
CCC1=CC=CC=C1NC(=O)CCCCl
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
42276-57-3
C12H16ClNO
225.716
MFCD03362124
VAWRRNXJFXHSSI-UHFFFAOYSA-N
4-chloro-n-2-ethylphenyl butanamide, 4-chloro-n-2-ethylphenyl butyramide
3523909
4-Chlor-N-(2-ethylphenyl)butanamid
CCC1=CC=CC=C1NC(=O)CCCCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-N-(2-ethylphenyl)butyramide
Menge 250 mg
Formelmasse 225.71
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'