missing translation for 'onlineSavingsMsg'
Learn More

4-Chlor-N-Cyclohexyl-N-Methyl-3-Nitrobenzamid, 97 %, Thermo Scientific™

Catalog Number p-7039847
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41614-18-0
C14H17ClN2O3
296.75
MFCD01211935
YPYXOOOTWPRAEX-UHFFFAOYSA-N
4-chloro-3-nitrophenyl-n-cyclohexyl-n-methylcarboxamide
3405764
4-chloro-N-cyclohexyl-N-methyl-3-nitrobenzamide
CN(C1CCCCC1)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
41614-18-0
C14H17ClN2O3
296.75
MFCD01211935
YPYXOOOTWPRAEX-UHFFFAOYSA-N
4-chloro-3-nitrophenyl-n-cyclohexyl-n-methylcarboxamide
3405764
4-chloro-N-cyclohexyl-N-methyl-3-nitrobenzamide
CN(C1CCCCC1)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloro-N-cyclohexyl-N-methyl-3-nitrobenzamide
Menge 250 mg
Formelmasse 296.75
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'