missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobutyric anhydride, TRC

Artikelnummer. 30366122
Change view
Click to view available options
Menge:
250 mg
1 g
2.5 g
Packungsgröße:
1 Gramm
2.50 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30366122 2.5 g 2.50 Gramm
30356409 1 g 1 Gramm
30360880 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30366122 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC2929552.5G

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20496-01-9
Summenformel C8H12Cl2O3
Molekulargewicht (g/mol) 227.09
Synonym Butanoic acid, 4-chloro-, anhydride (9CI), Butyric acid, 4-chloro-, anhydride (7CI,8CI), 4-Chlorobutanoic anhydride
IUPAC-Name 4-chlorobutanoyl 4-chlorobutanoate
SMILES ClCCCC(=O)OC(=O)CCCCl

Specifications

Chemischer Name oder Material 4-Chlorobutyric anhydride
Menge 2.5 g
InChI-Formel InChI=1S/C8H12Cl2O3/c9-5-1-3-7(11)13-8(12)4-2-6-10/h1-6H2
Formelmasse 226.0163
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.