missing translation for 'onlineSavingsMsg'
Learn More

4-Chloromethyl-N-(4-fluorobenzyl)Benzamid, 97 %, Thermo Scientific™

Catalog Number p-7040367
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1094362-88-5
C15H13ClFNO
277.723
MFCD12568403
IWRWVLGYZZVJJR-UHFFFAOYSA-N
4-chloromethyl-n-4-fluorophenyl methyl benzamide, 4-chloromethyl-n-4-fluorobenzyl benzamide
43156288
4-(Chlormethyl)-N-[(4-fluorphenyl)methyl]benzamid
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)CCl)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1094362-88-5
C15H13ClFNO
277.723
MFCD12568403
IWRWVLGYZZVJJR-UHFFFAOYSA-N
4-chloromethyl-n-4-fluorophenyl methyl benzamide, 4-chloromethyl-n-4-fluorobenzyl benzamide
43156288
4-(Chlormethyl)-N-[(4-fluorphenyl)methyl]benzamid
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)CCl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Chloromethyl-N-(4-fluorobenzyl)benzamide
Menge 250 mg
Formelmasse 277.72
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'