missing translation for 'onlineSavingsMsg'
Learn More

4-[Difluor(3,4,5-trifluorphenoxy)methyl]-3,5-difluor-4 '-n-propylbiphenyl, 99 %, Thermo Scientific™

Catalog Number p-7037812
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
303186-20-1
C22H15F7O
428.35
MFCD18451990
YJUHXJQZJUQYJJ-UHFFFAOYSA-N
4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propyl-1,1'-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propylbiphenyl, 4-3,4,5-trifluorophenoxydifluoromethyl-3,5-difluoro-4'-propylbiphenyl, 2-difluoro-3,4,5-trifluorophenoxy methyl-1,3-difluoro-5-4-pro pylphenyl benzene, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4-propyl-1,1-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-n-propylbiphenyl
20654145
2-[difluor-(3,4,5-trifluorophenoxy)methyl]-1,3-difluor-5-(4-propylphenyl)benzol
CCCC1=CC=C(C=C1)C2=CC(=C(C(=C2)F)C(OC3=CC(=C(C(=C3)F)F)F)(F)F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
303186-20-1
C22H15F7O
428.35
MFCD18451990
YJUHXJQZJUQYJJ-UHFFFAOYSA-N
4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propyl-1,1'-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-propylbiphenyl, 4-3,4,5-trifluorophenoxydifluoromethyl-3,5-difluoro-4'-propylbiphenyl, 2-difluoro-3,4,5-trifluorophenoxy methyl-1,3-difluoro-5-4-pro pylphenyl benzene, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4-propyl-1,1-biphenyl, 4-difluoro 3,4,5-trifluorophenoxy methyl-3,5-difluoro-4'-n-propylbiphenyl
20654145
2-[difluor-(3,4,5-trifluorophenoxy)methyl]-1,3-difluor-5-(4-propylphenyl)benzol
CCCC1=CC=C(C=C1)C2=CC(=C(C(=C2)F)C(OC3=CC(=C(C(=C3)F)F)F)(F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-[Difluoro(3,4,5-trifluorophenoxy)methyl]-3,5-difluoro-4'-n-propylbiphenyl
Menge 1 g
Formelmasse 428.35
Reinheit (%) 99%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'