missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-3-(Trifluormethyl)phenol, 97 %

Catalog Number 15466497
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1206593-24-9
C9H9F3O2
206.164
MFCD13196587
KTAPXNQLDCIIAO-UHFFFAOYSA-N
4-ethoxy-3-trifluoromethyl phenol, 3-trifluoromethyl phenetyl alcohol
57361413
4-ethoxy-3-(trifluormethyl)phenol
CCOC1=C(C=C(C=C1)O)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
1206593-24-9
C9H9F3O2
206.164
MFCD13196587
KTAPXNQLDCIIAO-UHFFFAOYSA-N
4-ethoxy-3-trifluoromethyl phenol, 3-trifluoromethyl phenetyl alcohol
57361413
4-ethoxy-3-(trifluormethyl)phenol
CCOC1=C(C=C(C=C1)O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Ethoxy-3-(trifluoromethyl)phenol
Menge 1 g
Formelmasse 206.16
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'