missing translation for 'onlineSavingsMsg'
Learn More

4-Fluor-3-(2-hydroxyethylcarbamoyl)benzolboronsäure, 98 %, Thermo Scientific™

Catalog Number p-7035220
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874219-25-7
C9H11BFNO4
227.00
MFCD08235044
AGNPGTVSVSPBEX-UHFFFAOYSA-N
4-fluoro-3-2-hydroxyethyl carbamoyl phenyl boronic acid, 4-fluoro-3-2-hydroxyethylcarbamoyl phenylboronic acid, 5-borono-2-fluoro-n-2-hydroxyethyl benzamide, 4-fluoro-3-2-hydroxyethyl carbamoyl phenylboronic acid, acmc-209qkq, 4-fluoro-3-2-hydroxyethyl carbamoyl phenyl boronicacid, 4-fluoranyl-3-2-hydroxyethylcarbamoyl phenyl boronic acid, 4-fluoro-3-2-hydroxyethyl carbamoyl benzeneboronic acid
44717584
[4-fluor-3-(2-hydroxyethylcarbamoyl)phenyl]boronsäure
OCCNC(=O)C1=C(F)C=CC(=C1)B(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
874219-25-7
C9H11BFNO4
227.00
MFCD08235044
AGNPGTVSVSPBEX-UHFFFAOYSA-N
4-fluoro-3-2-hydroxyethyl carbamoyl phenyl boronic acid, 4-fluoro-3-2-hydroxyethylcarbamoyl phenylboronic acid, 5-borono-2-fluoro-n-2-hydroxyethyl benzamide, 4-fluoro-3-2-hydroxyethyl carbamoyl phenylboronic acid, acmc-209qkq, 4-fluoro-3-2-hydroxyethyl carbamoyl phenyl boronicacid, 4-fluoranyl-3-2-hydroxyethylcarbamoyl phenyl boronic acid, 4-fluoro-3-2-hydroxyethyl carbamoyl benzeneboronic acid
44717584
[4-fluor-3-(2-hydroxyethylcarbamoyl)phenyl]boronsäure
OCCNC(=O)C1=C(F)C=CC(=C1)B(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Fluoro-3-(2-hydroxyethylcarbamoyl)benzeneboronic acid
Schmelzpunkt 89°C to 91°C
Menge 250 mg
Formelmasse 227
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'