Learn More
4-Fluor-N-(4-fluorbenzyl)benzamid, 97 %, Thermo Scientific™
Catalog Number p-7038701
Click to view available options
Menge:
250 mg
1 g
512181-73-6
C14H11F2NO
247.245
CJFYBDORFMDXTG-UHFFFAOYSA-N
4-fluoro-n-4-fluorophenyl methyl benzamide, 4-fluoro-n-4-fluorobenzyl benzamide, n-4-fluorobenzyl-4-fluorobenzamide, 4-fluoro-n-4-fluoro-benzyl-benzamide
2303704
4-fluor-N-[(4-fluorphenyl)methyl]benzamid
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)F)F
512181-73-6
C14H11F2NO
247.245
CJFYBDORFMDXTG-UHFFFAOYSA-N
4-fluoro-n-4-fluorophenyl methyl benzamide, 4-fluoro-n-4-fluorobenzyl benzamide, n-4-fluorobenzyl-4-fluorobenzamide, 4-fluoro-n-4-fluoro-benzyl-benzamide
2303704
4-fluor-N-[(4-fluorphenyl)methyl]benzamid
C1=CC(=CC=C1CNC(=O)C2=CC=C(C=C2)F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Fluoro-N-(4-fluorobenzyl)benzamide
Menge 250 mg
Formelmasse 247.24
Reinheit (%) 97%