missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)-pyridin-2-carbonsäure, 95 %, Thermo Scientific Chemicals

Catalog Number p-7031742
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
923169-37-3
C7H7NO3
153.137
MFCD08447083
QMLYYPGJXCVEHA-UHFFFAOYSA-N
4-hydroxymethyl picolinic acid, 4-hydroxymethyl pyridine-2-carboxylic acid, 4-hydroxymethyl pyridine-2-carboxylicacid
22645434
4-(Hydroxymethyl)Pyridin-2-Carbonsäure
C1=CN=C(C=C1CO)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
923169-37-3
C7H7NO3
153.137
MFCD08447083
QMLYYPGJXCVEHA-UHFFFAOYSA-N
4-hydroxymethyl picolinic acid, 4-hydroxymethyl pyridine-2-carboxylic acid, 4-hydroxymethyl pyridine-2-carboxylicacid
22645434
4-(Hydroxymethyl)Pyridin-2-Carbonsäure
C1=CN=C(C=C1CO)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Hydroxymethyl)pyridine-2-carboxylic acid
Schmelzpunkt 178°C to 185°C (decomposition)
Menge 1 g
Formelmasse 153.15
Reinheit (%) 95%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'