missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(pentafluorthio)benzoesäure, 97 %, Thermo Scientific™

Catalog Number p-7029290
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
886502-37-0
C9H7F3O4
236.146
MFCD06660285
PDDVYCWKVQXOTR-UHFFFAOYSA-N
4-methoxy-2-trifluoromethoxy benzoic acid
17750721
4-methoxy-2-(trifluormethoxy)benzoesäure
COC1=CC(=C(C=C1)C(=O)O)OC(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
886502-37-0
C9H7F3O4
236.146
MFCD06660285
PDDVYCWKVQXOTR-UHFFFAOYSA-N
4-methoxy-2-trifluoromethoxy benzoic acid
17750721
4-methoxy-2-(trifluormethoxy)benzoesäure
COC1=CC(=C(C=C1)C(=O)O)OC(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methoxy-2-(trifluoromethoxy)benzoic acid
Schmelzpunkt 99°C to 101°C
Menge 1 g
Formelmasse 236.15
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'