missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-2-(trifluormethyl)-Benzolborsäure, Thermo Scientific Chemicals

Catalog Number p-7029584
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
313546-16-6
C8H8BF3O3
219.954
MFCD03095346
ZBCRZEJNAADYKG-UHFFFAOYSA-N
4-methoxy-2-trifluoromethyl phenylboronic acid, 4-methoxy-2-trifluoromethyl benzeneboronic acid, 4-methoxy-2-trifluoromethyl phenyl boronic acid, 4-methoxy-2-trifluoromethyl-phenylboronic acid, 4-methoxy-2, pubchem1868, acmc-1aefl, ablock ab-13-0076
12109460
[4-methoxy-2-(trifluormethyl)phenyl]boronsäure
B(C1=C(C=C(C=C1)OC)C(F)(F)F)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
313546-16-6
C8H8BF3O3
219.954
MFCD03095346
ZBCRZEJNAADYKG-UHFFFAOYSA-N
4-methoxy-2-trifluoromethyl phenylboronic acid, 4-methoxy-2-trifluoromethyl benzeneboronic acid, 4-methoxy-2-trifluoromethyl phenyl boronic acid, 4-methoxy-2-trifluoromethyl-phenylboronic acid, 4-methoxy-2, pubchem1868, acmc-1aefl, ablock ab-13-0076
12109460
[4-methoxy-2-(trifluormethyl)phenyl]boronsäure
B(C1=C(C=C(C=C1)OC)C(F)(F)F)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methoxy-2-(trifluoromethyl)benzeneboronic acid
Schmelzpunkt 168°C to 172°C
Menge 1 g
Formelmasse 219.95

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'