missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(4-morpholinylsulfonyl)benzenboronsäure, 95 %, Thermo Scientific™

Catalog Number 15440596
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871333-02-7
C11H16BNO6S
301.12
MFCD08056366
MCYOYCLLZQYKDW-UHFFFAOYSA-N
4-methoxy-3-morpholinosulfonyl phenyl boronic acid, 4-methoxy-3-n-morpholinylsulfonyl phenylboronic acid, 4-methoxy-3-morpholin-4-ylsulphonyl benzeneboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenylboronic acid, boronic acid,b-3-methoxy-5-4-morpholinylsulfonyl phenyl, acmc-209qhv, 4-borono-2-morpholin-4-yl sulphonyl anisole, 4-5-borono-2-methoxyphenyl sulphonyl morpholine, 4-methoxy-3-morpholinosulfonyl phenylboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenyl boronic acid
44119826
(4-methoxy-3-morpholin-4-ylsulfonylphenyl)boronsäure
B(C1=CC(=C(C=C1)OC)S(=O)(=O)N2CCOCC2)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
871333-02-7
C11H16BNO6S
301.12
MFCD08056366
MCYOYCLLZQYKDW-UHFFFAOYSA-N
4-methoxy-3-morpholinosulfonyl phenyl boronic acid, 4-methoxy-3-n-morpholinylsulfonyl phenylboronic acid, 4-methoxy-3-morpholin-4-ylsulphonyl benzeneboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenylboronic acid, boronic acid,b-3-methoxy-5-4-morpholinylsulfonyl phenyl, acmc-209qhv, 4-borono-2-morpholin-4-yl sulphonyl anisole, 4-5-borono-2-methoxyphenyl sulphonyl morpholine, 4-methoxy-3-morpholinosulfonyl phenylboronic acid, 4-methoxy-3-morpholine-4-sulfonyl phenyl boronic acid
44119826
(4-methoxy-3-morpholin-4-ylsulfonylphenyl)boronsäure
B(C1=CC(=C(C=C1)OC)S(=O)(=O)N2CCOCC2)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methoxy-3-(4-morpholinylsulfonyl)benzeneboronic acid
Schmelzpunkt 270°C to 276°C
Menge 250 mg
Formelmasse 301.12
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'