missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluormethyl)benzolsulfonylchlorid, 97 %, Thermo Scientific™

Catalog Number p-7031237
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
694514-21-1
C8H6ClF3O3S
274.638
MFCD07774167
AXNBCYIRTWOTHS-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl benzenesulphonyl chloride, 4-methoxy-3-trifluoromethyl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl
22482384
4-methoxy-3-(trifluormethyl)benzolsulfonychlorid
COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(F)(F)F
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
694514-21-1
C8H6ClF3O3S
274.638
MFCD07774167
AXNBCYIRTWOTHS-UHFFFAOYSA-N
4-methoxy-3-trifluoromethyl benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl benzenesulphonyl chloride, 4-methoxy-3-trifluoromethyl benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-methoxy-3-trifluoromethyl
22482384
4-methoxy-3-(trifluormethyl)benzolsulfonychlorid
COC1=C(C=C(C=C1)S(=O)(=O)Cl)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methoxy-3-(trifluoromethyl)benzenesulfonyl chloride
Schmelzpunkt 52°C to 55°C
Menge 250 mg
UN-Nummer UN3261
Empfindlichkeit Moisture sensitive
Formelmasse 274.64
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'