missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methoxybenzol-1,3-dicarbonsäure, Mikromol™
Beschreibung
4-Methoxybenzol-1,3-dicarbonsäure, Mikromol
Spezifikation
Spezifikation
| Chemischer Name oder Material | 4-methoxybenzene-1,3-dicarboxylic acid |
| CAS | 2206-43-1 |
| Bezeichnung der Analyten oder Bestandteile | 4-methoxybenzene-1,3-dicarboxylic acid |
| Versandbedingung | Room Temperature |
| Summenformel | C9 H8 O5 |
| Zertifizierung/Konformität | ISO 17025 |
| Synonym | Picotamide Monohydrate Imp. A (EP), Picotamide Imp. A (EP), 2-Methoxy-1,5-benzenedicarboxylic acid, 4-Methoxy-1,3-benzenedicarboxylic acid, 4-Methoxyisophthalic acid, NSC 20694, Picotamide Monohydrate Impurity A, Picotamide Impurity A |
| InChI-Formel | InChI=1S/C9H8O5/c1-14-7-3-2-5(8(10)11)4-6(7)9(12)13/h2-4H,1H3,(H,10,11)(H,12,13) |
| Lösungstyp | Neat |
| SMILES | COc1ccc(cc1C(=O)O)C(=O)O |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?