missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxybiphenyl-4-Carbonsäure, 95 %, Thermo Scientific™

Catalog Number p-7034587
Click to view available options
Menge:
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
725-14-4
C14H12O3
228.247
MFCD00185792
FDPKGXQCDURRBM-UHFFFAOYSA-N
4'-methoxybiphenyl-4-carboxylic acid, 4-4-methoxyphenyl benzoic acid, 4'-methoxy-1,1'-biphenyl-4-carboxylic acid, 4'-methoxy 1,1'-biphenyl-4-carboxylic acid, 4'-methoxy-biphenyl-4-carboxylic acid, 4'-methoxy-4-biphenylcarboxylic acid, 4-biphenyl-4'-methoxy carboxylic acid, 1,1'-biphenyl-4-carboxylicacid, 4'-methoxy, pubchem10293, acmc-1bc5q
2759552
4-(4-methoxyphenyl)benzoesäure
COC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
725-14-4
C14H12O3
228.247
MFCD00185792
FDPKGXQCDURRBM-UHFFFAOYSA-N
4'-methoxybiphenyl-4-carboxylic acid, 4-4-methoxyphenyl benzoic acid, 4'-methoxy-1,1'-biphenyl-4-carboxylic acid, 4'-methoxy 1,1'-biphenyl-4-carboxylic acid, 4'-methoxy-biphenyl-4-carboxylic acid, 4'-methoxy-4-biphenylcarboxylic acid, 4-biphenyl-4'-methoxy carboxylic acid, 1,1'-biphenyl-4-carboxylicacid, 4'-methoxy, pubchem10293, acmc-1bc5q
2759552
4-(4-methoxyphenyl)benzoesäure
COC1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4'-Methoxybiphenyl-4-carboxylic acid
Menge 1 g
Formelmasse 228.25
Reinheit (%) 95%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'