missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonylamino)Benzenboronsäure, 97 %, Thermo Scientific Chemicals

Catalog Number p-7035313
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850567-96-3
C8H10BNO4
194.981
MFCD03788425
ROAKVYQVTMOEGR-UHFFFAOYSA-N
4-methoxycarbonylamino phenylboronic acid, 4-methoxycarbonyl amino phenyl boronic acid, 4-methoxycarbonylamino benzeneboronic acid, 4-methoxycarbonyl amino phenylboronic acid, 4-methoxycarbonylamino phenyl boronic acid, acmc-209q0y, 4-methoxycarbonylamino-phenyl boronic acid, 4-methoxycarbonylamino benzene boronic acid, 4-methoxycarbonyl amino phenyl boronicacid
2773494
[4-(methoxycarbonylamino)phenyl]boronsäure
B(C1=CC=C(C=C1)NC(=O)OC)(O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
850567-96-3
C8H10BNO4
194.981
MFCD03788425
ROAKVYQVTMOEGR-UHFFFAOYSA-N
4-methoxycarbonylamino phenylboronic acid, 4-methoxycarbonyl amino phenyl boronic acid, 4-methoxycarbonylamino benzeneboronic acid, 4-methoxycarbonyl amino phenylboronic acid, 4-methoxycarbonylamino phenyl boronic acid, acmc-209q0y, 4-methoxycarbonylamino-phenyl boronic acid, 4-methoxycarbonylamino benzene boronic acid, 4-methoxycarbonyl amino phenyl boronicacid
2773494
[4-(methoxycarbonylamino)phenyl]boronsäure
B(C1=CC=C(C=C1)NC(=O)OC)(O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(Methoxycarbonylamino)benzeneboronic acid
Schmelzpunkt 234°C to 236°C
Menge 250 mg
Formelmasse 194.98
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'