missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-N-Phenylbenzamid, 97 %, Thermo Scientific™

Catalog Number p-7038166
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6833-18-7
C14H13NO
211.264
MFCD00017213
PQEOPHYIUYAVDQ-UHFFFAOYSA-N
730569
4-methyl-N-phenylbenzamid
CC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
6833-18-7
C14H13NO
211.264
MFCD00017213
PQEOPHYIUYAVDQ-UHFFFAOYSA-N
730569
4-methyl-N-phenylbenzamid
CC1=CC=C(C=C1)C(=O)NC2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methyl-N-phenylbenzamide
Menge 250 mg
Synonym 4-methylbenzanilide, benzamide, 4-methyl-n-phenyl, p-methyl benzanilide, benzamide,4-methyl-n-phenyl, cbmicro_015159, n-phenyl-4-methylbenzamide, 4-methylphenyl-n-benzamide
Formelmasse 211.26
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'