missing translation for 'onlineSavingsMsg'
Learn More

4-Methylanisol, 99 %, Thermo Scientific Chemicals

Catalog Number p-7081555
Click to view available options
Menge:
100 mL
500 mL
2500 mL
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
104-93-8
C8H10O
122.167
MFCD00008413
CHLICZRVGGXEOD-UHFFFAOYSA-N
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
7731
1-Methoxy-4-methylbenzol
CC1=CC=C(C=C1)OC
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
104-93-8
C8H10O
122.167
MFCD00008413
CHLICZRVGGXEOD-UHFFFAOYSA-N
4-methylanisole, p-methylanisole, 4-methoxytoluene, p-methoxytoluene, benzene, 1-methoxy-4-methyl, p-cresyl methyl ether, p-cresol methyl ether, anisole, p-methyl, methyl p-tolyl ether, p-tolyl methyl ether
7731
1-Methoxy-4-methylbenzol
CC1=CC=C(C=C1)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Methylanisole
Schmelzpunkt -50°C
Siedepunkt 174°C to 175°C
Flammpunkt 58°C (136°F)
Geruch Aromatic
Menge 100 mL
UN-Nummer UN3271
Beilstein 1237336
Formelmasse 122.17
Reinheit (%) 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'