missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 4-Methylhistamine dihydrochloride
Selective, high affinity H4 agonist
N.V.
Spezifikation
Chemischer Name oder Material | 5-(2-Aminoethyl)-4-methylimidazole dihydrochloride |
---|---|
CAS | 36376-47-3 |
Löslichkeit | Soluble to 50mM in water |
Summenformel | C6H11N3.2HCl |
Formelmasse | Observed MW: 198.09 |
Beschreibung
Potent, high affinity H4 receptor agonist (Ki = 7 nM) that displays > 100-fold selectivity over other human histamine receptor subtypes.Spezifikation
5-(2-Aminoethyl)-4-methylimidazole dihydrochloride | |
Soluble to 50mM in water | |
Observed MW: 198.09 |
36376-47-3 | |
C6H11N3.2HCl |
For Research Use Only