Learn More
Alfa Aesar™ 4-n-Butoxyanilin, 97 %
Marke: Alfa Aesar™ A17528.14
Weitere Details : CAS : 4344-55-2 Netto-Gewicht : 0.02500kg Transport : UN number : 2810 Chem class : 6.1 Pack group : III
Menge | 25g |
---|
Chemische Identifikatoren
4344-55-2 | |
165.236 | |
UBRIHZOFEJHMIT-UHFFFAOYSA-N | |
20352 | |
CCCCOC1=CC=C(C=C1)N |
C10H15NO | |
MFCD00007866 | |
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline | |
4-Butoxyanilin |
Spezifikation
4-n-Butoxyaniline | |
>110°C (230°F) | |
84°C to 86°C (0.05mmHg) | |
4344-55-2 | |
MFCD00007866 | |
2084421 | |
UBRIHZOFEJHMIT-UHFFFAOYSA-N | |
4-Butoxyanilin | |
20352 | |
97% |
0.992 | |
1.538 | |
25g | |
C10H15NO | |
UN2810 | |
p-butoxyaniline, benzenamine, 4-butoxy, p-butyloxyaniline, 4-n-butoxyaniline, aniline, p-butoxy, p-aminophenol n-butyl ether, 4-butyloxybenzenamine, aminophenyl-n-butyl ether, 4-butyloxyaniline, 4-butyloxy aniline | |
CCCCOC1=CC=C(C=C1)N | |
165.236 | |
165.24 |