missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Undecyloxy)benzoesäure, 98 %, Thermo Scientific™

Catalog Number p-7027736
Click to view available options
Menge:
1 g
5 g
25 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
15872-44-3
C18H28O3
292.419
MFCD00043772
NEJZHJHZOUISSH-UHFFFAOYSA-N
4-undecyloxy benzoic acid, 4-undecyloxybenzoic acid, 4-n-undecyloxybenzoic acid, p-undecyloxybenzoic acid, pubchem3929, 4-undecyloxybenzoicacid, acmc-209diw, 4-undecyloxy benzoic acid #, benzoic acid,4-undecyloxy, timtec-bb sbb008333
519187
4-Undecoxybenzoesäure
CCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
15872-44-3
C18H28O3
292.419
MFCD00043772
NEJZHJHZOUISSH-UHFFFAOYSA-N
4-undecyloxy benzoic acid, 4-undecyloxybenzoic acid, 4-n-undecyloxybenzoic acid, p-undecyloxybenzoic acid, pubchem3929, 4-undecyloxybenzoicacid, acmc-209diw, 4-undecyloxy benzoic acid #, benzoic acid,4-undecyloxy, timtec-bb sbb008333
519187
4-Undecoxybenzoesäure
CCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-(n-Undecyloxy)benzoic acid
Siedepunkt 96°C to 137°C
Menge 1 g
Formelmasse 292.42
Reinheit (%) 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'