missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro Apremilast, TRC

Product Code. 30479536
Change view
Click to view available options
Menge:
250 mg
Unit Size:
250 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30479536 250 mg 250 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30479536 Supplier Toronto Research Chemicals Supplier No. TRCN598330250MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material 4-Nitro Apremilast
CAS 2095036-40-9
Summenformel C20 H20 N2 O8 S
Menge 250 mg
Synonym 1H-Isoindole-1,3(2H)-dione','2-[(1S)-1-(3-ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-4-nitro- (ACI), 2-[(1S)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethyl]-4-nitro-1H-isoindole-1,3(2H)-dione (ACI), (S)-2-[1-(3-Ethoxy-4-methoxyphenyl)-2-(methyl
InChI-Formel InChI=1S/C20H20N2O8S/c1-4-30-17-10-12(8-9-16(17)29-2)15(11-31(3,27)28)21-19(23)13-6-5-7-14(22(25)26)18(13)20(21)24/h5-10,15H,4,11H2,1-3H3/t15-/m1/s1
SMILES CCOc1cc(ccc1OC)[C@@H](CS(=O)(=O)C)N2C(=O)c3cccc(c3C2=O)[N+](=O)[O-]
IUPAC-Name 2-[(1S)-1-(3-Ethoxy-4-methoxyphenyl)-2-methylsulfonylethyl]-4-nitroisoindol-1,3-dion
Molekulargewicht (g/mol) 448.446
Formelmasse 448.094
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.