missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenyl-α-D-glucopyranosid, 99 %, Thermo Scientific Chemicals

Catalog Number p-3737073
Click to view available options
Menge:
1 g
5 g
25 g
Verpackung:
Glasflasche
3767-28-0
C12H15NO8
301.25
MFCD00064088
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside
92969
(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-Nitrophenoxy)oxan-3,4,5-Triol
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O
3767-28-0
C12H15NO8
301.25
MFCD00064088
IFBHRQDFSNCLOZ-ZIQFBCGOSA-N
4-nitrophenyl-alpha-d-glucopyranoside, 4-nitrophenyl alpha-d-glucopyranoside, 4-nitrophenyl a-d-glucopyranoside, 4-nitrophenyl alpha-glucoside, pnpalphaglu, 2r,3s,4s,5r,6r-2-hydroxymethyl-6-4-nitrophenoxy oxane-3,4,5-triol, p-nitrophenyl alpha-d-glucopyranoside, 4'-nitrophenyl-alpha-d-glucopyranoside, png, p-nitrophenyl, a-d-glucopyranoside
92969
(2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-Nitrophenoxy)oxan-3,4,5-Triol
C1=CC(=CC=C1[N+](=O)[O-])OC2C(C(C(C(O2)CO)O)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 4-Nitrophenyl-α-D-glucopyranoside
CAS Min. % 98.5
CAS Max. % 100.0
Schmelzpunkt 204.0°C to 218.0°C
Farbe Weiß bis gelb
Infrarotspektrum Echt
Prozentgehaltsbereich 98.5% min. (HPLC)
Verpackung Glasflasche
Menge 1 g
Spezifischer Drehzustand +218.00 (20.00°C c=0.5,water)
Spezifische Drehung + 218.00
Löslichkeitsinformationen Colorless to green-yellow solution
Formelmasse 301.25
Reinheit (%) 99%
Physikalische Form Kristallpulver
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'