Learn More
4-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 263021-30-3 |
| Molecular Formula | C13H17NO4 |
| MDL Number | MFCD04972273 |
| Quantity | 25 g |
| SMILES | O=C(O)C1=CC=C(N(C(OC(C)(C)C)=O)C)C=C1 |
| Molecular Weight (g/mol) | 251.28 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid |